2-Thienylacetyl chloride ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Thiopheneacetyl chloride, Thiophene-2-acetyl chloride

114508-1G 114508-10G
102894-188EA 60 USD
102894-188 102894-190
2-Thienylacetyl chloride ≥97%
2-Thienylacetyl chloride

Hazard: Irritant

Formula: C₆H₅ClOS
MW: 160.62 g/mol
MDL Number: MFCD00005456
CAS Number: 39098-97-0
UN: 3265
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR