6-Morpholinonicotinoyl chloride 95
Supplier: Matrix Scientific
114481-1G
114481-10G
102894-132EA
170.77
USD
102894-132
102894-134
6-Morpholinonicotinoyl chloride 95
6-Morpholinonicotinoyl chloride
Hazard: Irritant
Formula:
C₁₀H₁₁ClN₂O₂ MW: 226.66 g/mol |
MDL Number:
MFCD02681966 CAS Number: 313350-36-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...