3-(Diethylenetriamino)propylfunctionalised silica gel ≥97%

Supplier: Matrix Scientific

Synonyms: Triamine-3 functionalized silica gel

114391-1G 114391-10G 114391-50G
102893-932EA 109.23 USD
102893-932 102893-934 102893-936
3-(Diethylenetriamino)propylfunctionalised silica gel ≥97%
3-(Diethylenetriamino)propylfunctionalised silica gel

Hazard: Irritant

Formula: C₇H₁₉N₃OSi
MW: 189.33 g/mol
CAS Number: 1173022-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR