2-Methoxy-5-(trifluoromethoxy)aniline ≥97%
Supplier: Matrix Scientific
114370-1G
114370-10G
102893-888EA
109.23
USD
102893-888
102893-890
2-Methoxy-5-(trifluoromethoxy)aniline ≥97%
2-Methoxy-5-(trifluoromethoxy)aniline
Hazard: Irritant
Formula:
C₈H₈F₃NO₂ |
MDL Number:
MFCD09832305 CAS Number: 660848-57-7 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...