(4-Bromophenyl)acetyl chloride ≥97%

Supplier: Matrix Scientific

Synonyms: 2-(4-Bromophenyl)acetyl chloride

114336-1G 114336-10G
102893-818EA 156.92 USD
102893-818 102893-820
(4-Bromophenyl)acetyl chloride ≥97%
(4-Bromophenyl)acetyl chloride

Hazard: Irritant

Formula: C₈H₆BrClO
MW: 233.5 g/mol
MDL Number: MFCD00126842
CAS Number: 37859-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR