1,1,3,3-Tetramethylbutyl isocyanide ≥97%

Supplier: Matrix Scientific

Synonyms: 1,1,3,3-Tetramethylbutylisocyanide

114274-1G 114274-10G
102893-708EA 90.77 USD
102893-708 102893-710
1,1,3,3-Tetramethylbutyl isocyanide ≥97%
1,1,3,3-Tetramethylbutyl isocyanide

Hazard: Flammable

Formula: C₉H₁₇N
MW: 139.2380 g/mol
Boiling Pt: 50 °C (1.6 kPa)
Density: 0.8
Flash Pt: 52 °C
MDL Number: MFCD00000003
CAS Number: 14542-93-9
UN: 3275
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR