Thieno[2,3-d]pyrimidine-2,4-diol ≥97%

Supplier: Matrix Scientific

114256-1G 114256-10G
102893-664EA 241.54 USD
102893-664 102893-666
Thieno[2,3-d]pyrimidine-2,4-diol ≥97%
Thieno[2,3-d]pyrimidine-2,4-diol

Hazard: Irritant

Formula: C₆H₄N₂O₂S
MDL Number: MFCD11520884
CAS Number: 18740-38-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR