2-Mercaptopyridine-N-oxide ≥97%
Supplier: Matrix Scientific
Synonyms:
2-Mercaptopyridine 1-oxide
114203-1G
114203-10G
102893-550EA
109.23
USD
102893-550
102893-552
2-Mercaptopyridine-N-oxide ≥97%
2-Mercaptopyridine N-oxide
Hazard: Irritant
Formula:
C₅H₅NOS MW: 127.17 g/mol |
MDL Number:
MFCD00006196 CAS Number: 1121-31-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...