2-Chloro-6-fluorobenzaldehyde oxime ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-6-fluorobenzaldoxime

114156-1G 114156-10G
102893-462EA 86.15 USD
102893-462 102893-464
2-Chloro-6-fluorobenzaldehyde oxime ≥97%
2-Chloro-6-fluorobenzaldehyde oxime

Hazard: Irritant

Formula: C₇H₅ClFNO
MW: 173.57 g/mol
MDL Number: MFCD00837704
CAS Number: 443-33-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR