2'-Methyl-4'-nitroacetanilide ≥97%

Supplier: Matrix Scientific

114155-1G 114155-10G
102893-458EA 109.23 USD
102893-458 102893-460
2'-Methyl-4'-nitroacetanilide ≥97%
2'-Methyl-4'-nitroacetanilide

Hazard: Irritant

Formula: C₉H₁₀N₂O₃
MW: 194.19 g/mol
MDL Number: MFCD00033896
CAS Number: 2719-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR