2-Ethylpiperidine ≥97%

Supplier: Matrix Scientific

114133-1G 114133-10G
102893-408EA 60 USD
102893-408 102893-410
2-Ethylpiperidine ≥97%
2-Ethylpiperidine

Hazard: Flammable/Hygroscopic

Formula: C₇H₁₅N
MW: 113.2 g/mol
Boiling Pt: 143 °C
Density: 0.85
Flash Pt: 31 °C
MDL Number: MFCD00005988
CAS Number: 1484-80-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR