8-Nitroquinaldine ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Methyl-8-nitroquinoline

114119-1G 114119-10G
102893-350EA 86.15 USD
102893-350 102893-352
8-Nitroquinaldine ≥97%
8-Nitroquinaldine

Hazard: Irritant

Formula: C₁₀H₈N₂O₂
MW: 188.19 g/mol
MDL Number: MFCD00006764
CAS Number: 881-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR