5-Nitro-1,3-dihydro-2H-benzimidazol-2-one ≥97%

Supplier: Matrix Scientific

114117-1G 114117-10G
102893-344EA 60 USD
102893-344 102893-346
5-Nitro-1,3-dihydro-2H-benzimidazol-2-one ≥97%
5-Nitro-1,3-dihydro-2H-benzimidazol-2-one

Hazard: Irritant

Formula: C₇H₅N₃O₃
MDL Number: MFCD00220274
CAS Number: 93-84-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR