Ethyl-2-amino-4-hydroxypyrimidine-5-carboxylate ≥97%

Supplier: Matrix Scientific

114000-1G 114000-10G
102893-108EA 109.23 USD
102893-108 102893-110
Ethyl-2-amino-4-hydroxypyrimidine-5-carboxylate ≥97%
Ethyl-2-amino-4-hydroxypyrimidine-5-carboxylate

Hazard: Irritant

Formula: C₇H₉N₃O₃
MDL Number: MFCD00039708
CAS Number: 15400-53-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR