Ethyl-2-chloro-4-methyl-1,3-thiazole-5-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-chloro-4-methylthiazole-5-carboxylate

113993-1G 113993-10G
102893-092EA 329.23 USD
102893-092 102893-094
Ethyl-2-chloro-4-methyl-1,3-thiazole-5-carboxylate ≥97%
Ethyl-2-chloro-4-methyl-1,3-thiazole-5-carboxylate

Hazard: Irritant

Formula: C₇H₈ClNO₂S
MW: 205.66 g/mol
MDL Number: MFCD02956798
CAS Number: 7238-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR