Dimethyl adipate ≥97%

Supplier: Matrix Scientific

Synonyms: Adipic acid dimethyl ester, 1,6-Hexanedioic acid dimethyl ester

113982-10G 113982-50G
102893-084EA 41.54 USD
102893-084 102893-086
Dimethyl adipate ≥97%
Dimethyl adipate

Hazard: Irritant

Formula: C₈H₁₄O₄
MW: 174.2 g/mol
Boiling Pt: 229
Melting Pt: 10.3
Flash Pt: 107
MDL Number: MFCD00008469
CAS Number: 627-93-0
Merck Index: 13,00163

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR