Ethyl-5-amino-4-cyano-3-(2-ethoxycarbonylmethyl)thiophene-2-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Amino-4-cyano-3-(2-ethoxycarbonylmethyl)thiophene-2-carboxylic Acid Ethyl Ester, Ethyl 5-amino-4-cyano-3-(2-ethoxy-2-oxoethyl)thiophene-2-carboxylate

113972-1G 113972-10G
102898-338EA 156.92 USD
102898-338 102898-340
Ethyl-5-amino-4-cyano-3-(2-ethoxycarbonylmethyl)thiophene-2-carboxylate ≥97%
Ethyl-5-amino-4-cyano-3-(2-ethoxycarbonylmethyl)thiophene-2-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₄N₂O₄S
MW: 282.31 g/mol
MDL Number: MFCD00832845
CAS Number: 58168-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR