4-Methylcyclohexanecarboxylic acid (cis and trans mixture) ≥97%

Supplier: Matrix Scientific

113906-10G 113906-1G
102892-970EA 1333.85 USD
102892-970 102898-246
4-Methylcyclohexanecarboxylic acid (cis and trans mixture) ≥97%
4-Methylcyclohexanecarboxylic acid (cis and trans mixture)

Hazard: Irritant

Formula: C₈H₁₄O₂
MDL Number: MFCD00074909
CAS Number: 4331-54-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR