Imidazole-4,5-dicarboxylic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 1H-Imidazole-4,5-dicarboxylic acid, 4,5-Imidazoledicarboxylic acid

113841-1G 113841-10G 113841-50G
102892-894EA 51 USD
102892-894 102892-896 102892-898
Imidazole-4,5-dicarboxylic acid ≥97%
Imidazole-4,5-dicarboxylic acid

Hazard: Irritant

Formula: C₅H₄N₂O₄
MW: 156.10 g/mol
MDL Number: MFCD00005200
CAS Number: 570-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR