4,5-Difluorophthalonitrile ≥95%

Supplier: Matrix Scientific

113751-1G 113751-10G
102898-028EA 170.77 USD
102898-028 102898-030
4,5-Difluorophthalonitrile ≥95%
4,5-Difluorophthalonitrile

Hazard: Irritant

Formula: C₈H₂F₂N₂
MDL Number: MFCD00061281
CAS Number: 134450-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR