N-(Benzyloxycarbonyloxy)succinimide ≥97%
Supplier: Matrix Scientific
Synonyms:
CBZ-OSu (Z-OSu)
113743-1G
113743-10G
102892-730EA
38.46
USD
102892-730
102892-732
N-(Benzyloxycarbonyloxy)succinimide ≥97%
N-(Benzyloxycarbonyloxy)succinimide
Hazard: Irritant
Formula:
C₁₂H₁₁NO₅ MW: 249.22 g/mol |
MDL Number:
MFCD00005513 CAS Number: 13139-17-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...