Glutaric anhydride ≥97%
Supplier: Matrix Scientific
Synonyms:
Dihydro-2H-pyran-2,6(3H)-dione, Dihydro-2H-pyran-2,6(3H)-dione
113705-10G
113705-50G
102898-000EA
67.69
USD
102898-000
102898-002
Glutaric anhydride ≥97%
Glutaric anhydride
Hazard: Irritant
Formula:
C₅H₆O₃ MW: 114.1 g/mol |
MDL Number:
MFCD00006679 CAS Number: 108-55-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...