2-Ethoxy-1-naphthoic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Ethoxynaphthoic acid

113650-1G 113650-10G
102897-974EA 49.23 USD
102897-974 102897-976
2-Ethoxy-1-naphthoic acid ≥97%
2-Ethoxy-1-naphthoic acid

Hazard: Irritant

Formula: C₁₃H₁₂O₃
MW: 216.24 g/mol
MDL Number: MFCD00004008
CAS Number: 2224-00-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR