Atropic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Phenylacrylic Acid, Ipratropium bromide Impurity D

113624-1G 113624-10G
102897-924EA 364.62 USD
102897-924 102897-926
Atropic acid ≥97%
Atropic acid

Hazard: Irritant

Formula: C₉H₈O₂
MW: 148.16 g/mol
MDL Number: MFCD00046531
CAS Number: 492-38-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR