3-Amino-4-bromobenzoic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Amino-4-bromobenzoic Acid

113619-1G 113619-10G 113619-50G
102897-910EA 86.15 USD
102897-910 102897-912 102897-914
3-Amino-4-bromobenzoic acid ≥97%
3-Amino-4-bromobenzoic acid

Hazard: Irritant

Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
MDL Number: MFCD00579106
CAS Number: 2840-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR