2-(Difluoromethoxy)benzoic acid ≥97%

Supplier: Matrix Scientific

113615-1G 113615-10G
102897-900EA 73.85 USD
102897-900 102897-902
2-(Difluoromethoxy)benzoic acid ≥97%
2-(Difluoromethoxy)benzoic acid

Hazard: Irritant

Formula: C₈H₆F₂O₃
MDL Number: MFCD00236223
CAS Number: 97914-59-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR