2,4,6-Trimethylbenzaldehyde ≥97%
Supplier: Matrix Scientific
Synonyms:
Mesitaldehyde
113558-1G
113558-10G
102892-430EA
38.46
USD
102892-430
102892-432
2,4,6-Trimethylbenzaldehyde ≥97%
2,4,6-Trimethylbenzaldehyde
Hazard: Irritant
Formula:
C₁₀H₁₂O MW: 148.2 g/mol |
MDL Number:
MFCD00003341 CAS Number: 487-68-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...