Neopentylamine ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Amino-2,2-dimethylpropane

113547-1G 113547-10G
102897-866EA 135.38 USD
102897-866 102897-868
Neopentylamine ≥95%
Neopentylamine

Hazard: Irritant

Formula: C₅H₁₃N
MW: 87.16 g/mol
MDL Number: MFCD00008134
CAS Number: 5813-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR