1-(3-Dimethylaminopropyl)piperazine ≥97%
Supplier: Matrix Scientific
Synonyms:
N,N-Dimethyl-3-(piperazin-1-yl)propan-1-amine
113535-1G
113535-10G
102892-376EA
73.85
USD
102892-376
102892-378
1-(3-Dimethylaminopropyl)piperazine ≥97%
1-(3-Dimethylaminopropyl)piperazine
Hazard: Irritant
Formula:
C₉H₂₁N₃ MW: 171.29 g/mol |
MDL Number:
MFCD00082601 CAS Number: 877-96-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...