N,N-Dimethyl-4-nitroaniline ≥97%

Supplier: Matrix Scientific

113515-1G 113515-10G
102892-308EA 60 USD
102892-308 102892-310
N,N-Dimethyl-4-nitroaniline ≥97%
N,N-Dimethyl-4-nitroaniline

Hazard: Irritant

Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
MDL Number: MFCD00014712
CAS Number: 100-23-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR