4-Bromo-2,6-dichloropyridine ≥97%

Supplier: Matrix Scientific

113482-1G 113482-5G
102892-236EA 106.15 USD
102892-236 102892-238
4-Bromo-2,6-dichloropyridine ≥97%
4-Bromo-2,6-dichloropyridine

Hazard: Irritant

Formula: C₅H₂BrCl₂N
MW: 226.8900 g/mol
Melting Pt: 95-102°C
MDL Number: MFCD11848646
CAS Number: 98027-80-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR