5-Bromo-2-fluoroisonicotinic acid ≥98%

Supplier: Matrix Scientific

113476-250MG 113476-1G
102892-228EA 300 USD
102892-228 102892-230
5-Bromo-2-fluoroisonicotinic acid ≥98%
5-Bromo-2-fluoroisonicotinic acid

Hazard: Irritant

Formula: C₆H₃BrFNO₂
MDL Number: MFCD13185803
CAS Number: 1214377-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR