6-Bromo-5-fluoropyridine-2-carboxylic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 6-Bromo-5-fluoropicolinic acid

113475-250MG 113475-1G
102892-224EA 120 USD
102892-224 102892-226
6-Bromo-5-fluoropyridine-2-carboxylic acid ≥98%
6-Bromo-5-fluoropyridine-2-carboxylic acid

Hazard: Irritant

Formula: C₆H₃BrFNO₂
MW: 220 g/mol
MDL Number: MFCD13185797
CAS Number: 1052714-46-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR