cis-4-Aminocyclohexanemethanol hydrochloride
Supplier: Matrix Scientific
Synonyms:
(cis-4-Aminocyclohexyl)methanol hydrochloride
113470-1G
113470-5G
102897-860EA
727.69
USD
102897-860
102897-862
cis-4-Aminocyclohexanemethanol hydrochloride
cis-4-Aminocyclohexanemethanol hydrochloride
Hazard: Irritant
Formula:
C₇H₁₆ClNO MW: 165.66 g/mol |
MDL Number:
MFCD16661169 CAS Number: 61367-22-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...