4-(Boc-amino)-2-chloro-3-pyridinecarbaldehyde

Supplier: Matrix Scientific

Synonyms: tert-Butyl (2-chloro-3-formylpyridin-4-yl)carbamate

113455-1G 113455-5G
102892-206EA 727.69 USD
102892-206 102892-208
4-(Boc-amino)-2-chloro-3-pyridinecarbaldehyde
4-(Boc-amino)-2-chloro-3-pyridinecarbaldehyde

Hazard: Irritant

Formula: C₁₁H₁₃ClN₂O₃
MDL Number: MFCD16659013
CAS Number: 893423-62-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR