3,4-Pyridinedimethanol

Supplier: Matrix Scientific

113443-1G 113443-5G
102892-168EA 436.92 USD
102892-168 102892-200
3,4-Pyridinedimethanol
3,4-Pyridinedimethanol

Hazard: Irritant

Formula: C₇H₉NO₂
MDL Number: MFCD00160202
CAS Number: 38070-80-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR