2-Thiazolecarboximidamide hydrochloride

Supplier: Matrix Scientific

Synonyms: Thiazole-2-carboximidamide hydrochloride

113438-1G 113438-5G
102892-154EA 104.62 USD
102892-154 102892-156
2-Thiazolecarboximidamide hydrochloride
2-Thiazolecarboximidamide hydrochloride

Hazard: Irritant

Formula: C₄H₆ClN₃S
MW: 163.63 g/mol
MDL Number: MFCD18651714
CAS Number: 247037-82-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR