endo-3-(Boc-amino)-9-azabicyclo[3.3.1]nonane
Supplier: Matrix Scientific
113433-1G
113433-5G
102897-832EA
1047.69
USD
102897-832
102897-834
endo-3-(Boc-amino)-9-azabicyclo[3.3.1]nonane
endo-3-(Boc-amino)-9-azabicyclo[3.3.1]nonane
Hazard: Irritant
Formula:
C₁₃H₂₄N₂O₂ |
MDL Number:
MFCD19707046 CAS Number: 155560-04-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...