9-Benzyl-3-oxo-9-azabicyclo[3.3.1]nonane

Supplier: Matrix Scientific

113432-1G 113432-5G
102897-828EA 104.62 USD
102897-828 102897-830
9-Benzyl-3-oxo-9-azabicyclo[3.3.1]nonane
9-Benzyl-3-oxo-9-azabicyclo[3.3.1]nonane

Hazard: Irritant

Formula: C₁₅H₁₉NO
MDL Number: MFCD00144851
CAS Number: 2291-58-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR