9-Boc-3-oxo-9-azabicyclo[3.3.1]nonane

Supplier: Matrix Scientific

113431-1G 113431-5G
102897-824EA 170.77 USD
102897-824 102897-826
9-Boc-3-oxo-9-azabicyclo[3.3.1]nonane
9-Boc-3-oxo-9-azabicyclo[3.3.1]nonane

Hazard: Irritant

Formula: C₁₃H₂₁NO₃
MDL Number: MFCD09997838
CAS Number: 512822-27-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR