exo-3-Amino-9-methyl-9-azabicyclo[3.3.1]nonane

Supplier: Matrix Scientific

Synonyms: exo-3-Amino-9-methyl-9-azabicyclo[3,3,1]nonane

113430-1G 113430-5G
102897-820EA 581.54 USD
102897-820 102897-822
exo-3-Amino-9-methyl-9-azabicyclo[3.3.1]nonane
exo-3-Amino-9-methyl-9-azabicyclo[3.3.1]nonane

Hazard: Irritant

Formula: C₉H₁₈N₂
MW: 154.26 g/mol
MDL Number: MFCD12198832
CAS Number: 76272-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR