4,5-Dihydrofuran-3-carboxylic acid

Supplier: Matrix Scientific

113427-1G 113427-5G
102897-812EA 56.92 USD
102897-812 102897-814
4,5-Dihydrofuran-3-carboxylic acid
4,5-Dihydrofuran-3-carboxylic acid

Hazard: Irritant

Formula: C₅H₆O₃
MDL Number: MFCD07369946
CAS Number: 98021-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR