1-Isobutyl-1H-pyrazole-4-boronic acid pinacol ester

Supplier: Matrix Scientific

Synonyms: 1-Isobutyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 1-Isobutylpyrazole-4-boronic acid, pinacol ester

113420-1G 113420-5G
102892-142EA 170.77 USD
102892-142 102892-144
1-Isobutyl-1H-pyrazole-4-boronic acid pinacol ester
1-Isobutyl-1H-pyrazole-4-boronic acid pinacol ester

Hazard: Irritant

Formula: C₁₃H₂₃BN₂O₂
MW: 250.1449 g/mol
MDL Number: MFCD05663857
CAS Number: 827614-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR