Ethyl-2-formyl-3-oxopropionate

Supplier: Matrix Scientific

Synonyms: Ethyl 2-formyl-3-oxopropanoate, Ethyl 2-Formyl-3-oxopropionate, Diformylacetic acid ethyl ester, 2-Formyl-3-oxopropionic acid ethyl ester, Ethyl diformylacetate

113415-1G 113415-5G
102892-130EA 170.77 USD
102892-130 102892-132
Ethyl-2-formyl-3-oxopropionate
Ethyl-2-formyl-3-oxopropionate

Hazard: Irritant

Formula: C₆H₈O₄
MW: 144.13 g/mol
MDL Number: MFCD11112084
CAS Number: 80370-42-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR