5-Bromo-4-chloro-2-methylpyridine

Supplier: Matrix Scientific

113413-1G 113413-5G
102892-122EA 867.69 USD
102892-122 102892-124
5-Bromo-4-chloro-2-methylpyridine
5-Bromo-4-chloro-2-methylpyridine

Hazard: Irritant

Formula: C₆H₅BrClN
MW: 206.4700 g/mol
MDL Number: MFCD09839296
CAS Number: 1003711-85-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR