1,3-Dimethyl-6-amino-1H-indazole

Supplier: Matrix Scientific

Synonyms: 1,3-Dimethyl-1H-indazol-6-amine

113409-1G 113409-5G
102892-098EA 815.38 USD
102892-098 102892-100
1,3-Dimethyl-6-amino-1H-indazole
1,3-Dimethyl-6-amino-1H-indazole

Hazard: Irritant

Formula: C₉H₁₁N₃
MW: 161.21 g/mol
MDL Number: MFCD18432764
CAS Number: 221681-92-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR