3-Chloro-4-nitropyridine-1-oxide

Supplier: Matrix Scientific

Synonyms: 3-Chloro-4-nitropyridine N-oxide

113406-1G 113406-5G
102892-088EA 200 USD
102892-088 102892-090
3-Chloro-4-nitropyridine-1-oxide
3-Chloro-4-nitropyridine-1-oxide

Hazard: Irritant

Formula: C₅H₃ClN₂O₃
MW: 174.54 g/mol
MDL Number: MFCD00233948
CAS Number: 76439-45-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR