(R)-1-Methyl-2-aminomethylpyrrolidine

Supplier: Matrix Scientific

Synonyms: (R)-(1-Methylpyrrolidin-2-yl)methanamine

113400-1G 113400-5G
102897-756EA 1072.31 USD
102897-756 102897-758
(R)-1-Methyl-2-aminomethylpyrrolidine
(R)-1-Methyl-2-aminomethylpyrrolidine

Hazard: Irritant

Formula: C₆H₁₄N₂
MW: 114.19 g/mol
MDL Number: MFCD18425625
CAS Number: 66411-53-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR