trans-3-Aminocyclobutanol hydrochloride

Supplier: Matrix Scientific

113396-1G 113396-5G
102892-064EA 581.54 USD
102892-064 102892-066
trans-3-Aminocyclobutanol hydrochloride
trans-3-Aminocyclobutanol hydrochloride

Hazard: Irritant

Formula: C₄H₁₀ClNO
MDL Number: MFCD17214232
CAS Number: 1205037-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR