cis-tert-Butyl-3-hydroxycyclobutylcarbamate

Supplier: Matrix Scientific

Synonyms: tert-Butyl (cis-3-hydroxycyclobutyl)carbamate

113394-1G 113394-5G
102892-056EA 130.77 USD
102892-056 102892-058
cis-tert-Butyl-3-hydroxycyclobutylcarbamate
cis-tert-Butyl-3-hydroxycyclobutylcarbamate

Hazard: Irritant

Formula: C₉H₁₇NO₃
MW: 187.24 g/mol
MDL Number: MFCD09038208
CAS Number: 389890-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR